Cut resistant gloves, AP50
L, blauw/zwart
Catalogus # PRTWAP50B8RL
Leverancier: Portwest
Ga naar:
Product Details & Documents
The fully coated Aqua Cut Pro glove offers level D cut resistance.
- Sandy finish for exceptional grip in water, grease or oil
- Protects against cuts and abrasion
- Prevents grease, oil and water penetration
- Seamless 13 gauge liner
- Retail bag which aids presentation for retail sales
- ANSI Cut Level A4
The first coat is a smooth nitrile and the second coat is foam nitrile, providing a secure grip and a barrier against contaminants. Elastic wrist cuff provides a secure fit.
: AS/NZS 2161 .2, AS/NZS 2161 .3, ANSI/ISEA 105 - 2016 CUT Level (A4), EN 420, EN 388:2016 - (4.X.4.4.D)
Specificaties
- Handschoenmaat:L
- Kleur:blauw/zwart
- Seq:03
- Voeringmateriaal:HPPE
- Cat. No.:PRTWAP50B8RL
- Voor eenmalig gebruik/herbruikbaar:Herbruikbaar
- Manchetstijl:Elastisch
- Ambidexter/handspecifiek:Handspecifiek
- EN-norm:EN 420, EN 388:2016 - (4.X.4.4.D)
- Model:AP50
- Afwerking:Fully coated
- Steken per inch (Gauge):13



