DIN adapters for electrodes
DIN 19262 adapter T kit including temperature for LE438 / LE410
Catalogus # METT30499900
Leverancier: Mettler - Toledo
Ga naar:
Product Details & Documents
These adapters will allow the LE electrodes with BNC to connect to meters with DIN.
Specificaties
- Omschrijving:DIN 19262 adapter T kit including temperature for LE438 / LE410