[EN]MELANGE HF / HNO3 1 * 1 L
Catalogus # PRLS89962.290
Leverancier: PROLABO CMP
Frequently Bought Together
Natriumhydroxide 30% (w/w) in aqueous solution, GPR RECTAPUR®
Leverancier: VWR Chemicals
Teststrips, fosfaat, Reflectoquant®, Supelco®
Leverancier: Merck
Testsets, cyanidetest, voor Spectroquant®, Supelco®
Leverancier: Merck
pH-indicatiestrips, MQuant®, Supelco®
Leverancier: Merck
Immersion oil for microscopy, Sigma-Aldrich®
Leverancier: Merck
Teststrips, chromaattest MQuant™, Supelco®
Leverancier: Merck
L(+)-Glutamine, EMPROVE® EXPERT DAB, USP suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Kongorot, TECHNICAL
Leverancier: VWR Chemicals
Acid red 52, Technical Grade, Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
[EN]BIOTECTOR 1.8N SULFURIC ACID 80MG/L 1 * 20 L
Leverancier: Hach
NACL IN SOLUTION 1% +/-0.1% 1 * 100 mL
Leverancier: PROLABO CMP
Trypan blue 0,4% in aqueous solution, Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
Acid Red 183 30% (dye content), Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
Avantor® Seradigm, Fetal Bovine Serum (FBS), US origin
Leverancier: VWR Seradigm
Powder pillows, ammonia
Leverancier: Hach
Folin-Ciocalteu's reagent 1,9 - 2,1 N for analysis of phenols, suitable for determination of total protein by Lowry method, Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY