KIMAX ® volumetrische kolven met kleurcode [st] PTFE Stopper, brede mond, klasse A, 1 * 6 ST
KIML92812F-50
KIMAX ® volumetrische kolven met kleurcode [st] PTFE Stopper, brede mond, klasse A, 1 * 6 ST
KIML92812F-50
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, gezichtstissues
: VWR Collection
pH-teststrips, MQuant®, Supelco®
: Merck
VWR®, Nitrilhandschoenen
: VWR Collection
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Nitril handschoenen, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Serologische pipetten, standaardreeks
: VWR Collection
VWR®, Transferpipetten, Hoge Prestaties
: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandschoenen
: VWR Collection