Tool carts
Gereedschapswagen, met 5 laden, veiligheidsslot en cleanroomwielen met remmen, , 730×470×850 mm
Catalogus # PALBCT0702
Leverancier: Palbam
Ga naar:
Product Details & Documents
Tool cart in stainless steel 304 with electropolished finish.
- Five fully extendable drawers
- One lock which secures all the drawers
- Top ledge to prevent tools/parts falling
- Cleanroom castors with brakes
Specificaties
- Omschrijving:Gereedschapswagen, met 5 laden, veiligheidsslot en cleanroomwielen met remmen
- b×d×h:730×470×850 mm
- Cat. No.:PALBCT0702