Climbing helmets, PW97
Climbing helmet, PW97, wit
Catalogus # PRTWPW97WHR
Leverancier: Portwest
Ga naar:
Product Details & Documents
Lightweight and ventilated helmet for workers at height. High wear comfort and secure fit through ergonomically formed shell. The six point ratchet adjustable harness uses nylon suspension for wearer comfort.
- Y chin strap
- Molten metal
: EN 397 (–30/+50°C MM 440V), ANSI/ISEA Z89.1 Type I.
Specificaties
- Kleur:wit
- Omschrijving:Climbing helmet
- Seq:02
- Model:PW97



