Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Erlenmeyer flasks, PETG, with vented cap, Corning®
Erlenmeyer flasks, PETG, with vented cap, Corning®
2000 ml
Catalogus # CORN431280
Leverancier:   Corning
Erlenmeyer flasks, PETG, with vented cap, Corning®
Catalogus # CORN431280
Leverancier:  Corning
Erlenmeyer flasks, PETG, with vented cap, Corning®
Erlenmeyer flasks, PETG, with vented cap, Corning®
2000 ml
Price
282,00 €
Log in voor beschikbaarheid
1 * 6 ST
Product Details & Documents
PETG, optically clear, with vented cap.
  • Flasks will not break when dropped
  • Designed for single-use, as flasks will collapse if autoclaved
  • Molded-in graduations for accuracy
  • Individually packaged and radiation sterilised
  • Sterility Assurance Level (SAL) of 10⁻⁶
  • Nonpyrogenic
  • Baffled or plain bottom available in each size
These wide-mouth flasks are available in 2000, 3000 and 5000 ml sizes with either a baffled or plain bottom. The flasks are constructed of USP Class VI PETG for excellent optical clarity and mechanical strength. Molded graduations are standard, and threaded vent caps provide continuous gas exchange along with a liquid-tight seal. Vented threaded caps feature a 0,2 µm hydrophobic membrane to provide continuous gas exchange while ensuring sterility and preventing leakage.
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specificaties
  • Capaciteit:
    2000 ml
  • Seq:
    01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.