Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Cleaning cloths, MicroClean Plus

blauw, 400×450 mm, 5 cloths/pack
Catalogus # VILE152530
Leverancier:   FHP VILEDA SCS
Cleaning cloths, MicroClean Plus
Catalogus # VILE152530
Leverancier:  FHP VILEDA SCS
Cleaning cloths, MicroClean Plus
Cleaning cloths, MicroClean Plus

blauw, 400×450 mm, 5 cloths/pack

Price
621,00 €
Log in voor beschikbaarheid
1 * 100 ST
Product Details & Documents

MicroClean Plus is a high performance cleaning cloth made from a uniquely fine microfibre (70% polyester and 
30% polyamide). Great streak- and lint-free results are achieved with just one wipe. Ideal for quick dirt removal even without chemicals.

  • Extremly durable
  • Micron Fibre Technology for deeper and more effective cleaning
  • MicroClean dries quickly: Bacteria growth is prevented
  • Washable and reusable: Reduces waste and protects the environment
  • Colour coding system allows a cleaning process according to hygiene standards

MicroClean Plus is suitable for demanding cleaning tasks in general building cleaning and Hotels/Restaurants/Catering on every kind of surface - with or without cleaning agents.

Specificaties
  • Verpakt:
    5 cloths/pack
  • Kleur:
    blauw
  • l×b:
    400×450 mm