Kits for Precellys®
Precellys® Glass kit 0,5 mm, 25×15 ml tubes, pre-filled with glass beads, Micro-organismen, VK05/ 15 ml
Catalogus # 432-0350
Leverancier: Bertin Instruments
Ga naar:
Product Details & Documents
The Precellys lysing kits offer the best versatility of volumes and grinding material to process any type of samples: Human, animal, plants, microorganisms or soil.
- More than 30 different lysing matrixes
- Reinforced tubes are suitable for use at the maximum energy levels
- Easy-handling features allow the Precellys to be used without any advanced training
These kits are specifically for sample preparation for Precellys homogenisers. They can process any type of sample. Choosing the appropriate lysing kit maximises the quantity and quality of extraction by decreasing the process time and limiting the heat generated.
Reinforced tubes have thicker walls and their top contains an o-ring to make it more resistant.
Documents
Certificaten
Zoeken op lotnummerBertin-Precellys-Lysing-Kits Katalogseite Precellys Aufschluss-Kits
Product documentatie
BERTP000913-LYSK0A_240627-890.pdf BERTP000947-LYSK0A_240716-830.pdf CoC_BERTP000913-LYSK0A_200707-804 CoC_BERTP000913-LYSK0A_210809-890 coc_BERTP000916-LYSK0A_250716-830
More
Hair Grinding In 7 ml Tube Hard Sample Grinding with 7 ml Metal Tube Improving Compound Extraction Efficiency using the Precellys24 Protein Extraction From E.coli Using Precellys® Evolution Quantative recovery of BMP-2 after grinding of demineralised bone with Precellys® Evolution
More
Manual Precellys Emulsion Kit
Handmatig
Usage limits of Precellys lysing kits
Technische fiche
Specificaties
- Omschrijving:Precellys® Glass kit 0,5 mm, 25×15 ml tubes, pre-filled with glass beads
- Type:VK05/ 15 ml
- Voor:Micro-organismen