Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
High content cell imaging system, ImageXpress Micro 4
Filter set, DAPI
Catalog # MLDV5039856
Supplier:   Molecular Devices
High content cell imaging system, ImageXpress Micro 4
Catalog # MLDV5039856
Supplier:  Molecular Devices
High content cell imaging system, ImageXpress Micro 4
High content cell imaging system, ImageXpress Micro 4
Filter set, DAPI
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
The ImageXpress Micro 4 high content imaging system represents the culmination of four generations of imaging expertise. The agile design allows you to boost your research with this faster than ever system, while providing the option to upgrade to confocal in the future to align with your research needs.
  • Tackle a wide range of assays with oil and air objectives ranging from 1× to 100×
  • Automated five-position filter cube changer to facilitate multiplex assays with up to five filter sets and four objectives for a single experiment
  • Optically-encoded linear voice coil driven stages with encoders that measure position with better than 25 nm resolution, to achieve high accuracy and precision in X, Y and Z axes
  • Expand your research capabilities with environmental control: Temperature, humidity and CO₂ control for multi-day, live cell experiments), transmitted light true phase contrast, confocal imaging (field upgradable), and fluidics (onboard disposable pipetting)
Capture images of whole organisms and cellular or intracellular events with the ImageXpress Micro 4 system, configured to suit your specific biological needs. We offer unlimited configurability with user exchangeable filter cubes, a wide range of objective lenses and environmental control, transmitted light, confocal imaging and fluidic options. Speed up your research by acquiring more than 245000 wells per day at 4× magnification with the system’s unique multi-well crop feature, and resolve the fine detail you need with increasing magnifications.
Combined with MetaXpress high content image acquisition and analysis software, the ImageXpress Micro 4 system is an end-to-end solution to interpret images, understand data and explore ideas.
Specifications
  • Omschrijving:
    Filter set, DAPI
  • Seq:
    105
Complementary Products
Product Information
  • Catalog #
    Omschrijving
    VPE
    Serumformaat
    Steriliteit
    Verpakt
    Kleur
    Cultuuroppervlak
    Aanbevolen werkvolume
    Steriel
    Werkvolume
    Availability
    Price
  • 30026.
    Calcein-AM Cell Viability Assay kit, 1000 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 417,00 €
  • 30029.
    NucView™ 488 Caspase-3 Assay Kit for live cells, 100 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 631,00 €
  • S1810-500
    Foetaal runderserum, Zuid-Amerikaanse oorsprong
    Bevroren
    15
    Log in for availability
    1 * 500 mL - 330,00 €
  • 100.103.010
    GrowDex®, 10 ml syringe
    1
    Log in for availability
    1 * 1 ST - 290,00 €
  • 30002.
    Levensvatbaarheid/cytotoxiciteit-testkit voor dierlijke levende en dode cellen, 300 tests
    1 KIT
    Log in for availability
    1 * 1 KIT - 440,00 €
  • 30009-2
    Caspase-3 DEVD-R110 Fluorometric HTS Assay Kit, 10 ml
    1 KIT
    Log in for availability
    1 * 1 KIT - 294,00 €
  • 734-2327
    1/zak
    helder
    0,33 cm²
    0,075 - 0,20 ml
    +
    17
    Log in for availability
    1 * 100 ST - 238,00 €
  • 735-0523
    Microlite™ 1+ strip, 1×12 per strip
    Log in for availability
    1 * 320 ST - 231,00 €
  • CORN3904BC
    20/bag
    75 - 200 µl
    96-wells microplaat, heldere vlakke bodem, behandeld voor weefselcultuur, met deksel
    360 µl
    +
    21
    Log in for availability
    1 * 100 ST - 1 160,00 €
  • 734-2097
    transparant
    96-MicroWell™-plaat, wells met vlakke bodem, met deksel
    06
    Log in for availability
    1 * 50 ST - 197,00 €