D-(+)-Malic acid
Bottle, 25 g
APOSOR315008-25G
: (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid
- :636-61-3
- :C₄H₆O₅
- :MFCD00004245
- :134.09 g/mol
- :100…104 °C
- :1,6 g/cm³ (25 °C)
- :Kamertemperatuur
- :211-262-2
- Verpakking:Bottle
- VPE:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.