Orders LinkContactUsLinkComponent
D-(+)-Malic acid
Bottle, 25 g
  APOSOR315008-25G
D-(+)-Malic acid
  APOSOR315008-25G
 :  636-61-3
D-(+)-Malic acid
Bottle, 25 g
 :  636-61-3
27,50 €
1 * 25 g
 :  (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid
  •  :636-61-3
  •  :C₄H₆O₅
  •  :MFCD00004245
  •  :134.09 g/mol
  •  :100…104 °C
  •  :1,6 g/cm³ (25 °C)
  •  :Kamertemperatuur
  •  :211-262-2
  • Verpakking:
    Bottle
  • VPE:
    25 g