D-(+)-Malic acid
Bottle, 100 g
APOSOR315008-100G
: (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid
- :636-61-3
- :C₄H₆O₅
- :MFCD00004245
- :134.09 g/mol
- :100…104 °C
- :1,6 g/cm³ (25 °C)
- :Kamertemperatuur
- :211-262-2
- Verpakking:Bottle
- VPE:100 g