Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
4'-Fluoroacetophenone 99%

4'-Fluoroacetophenone 99%

Bottle, 1 kg
Catalogus # APOSPC3380-1KG
Irriterend
Leverancier:   Apollo Scientific
4'-Fluoroacetophenone 99%
Catalogus # APOSPC3380-1KG
Leverancier:  Apollo Scientific
CAS-nummer:  403-42-9
4'-Fluoroacetophenone 99%

Bottle, 1 kg

CAS-nummer:  403-42-9
Price
109,00 €
Log in voor beschikbaarheid
1 * 1 kg
Product Details & Documents
Synoniemen:  1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
  • CAS-nummer:403-42-9
  • Formule:C₈H₇FO
  • MDL-nummer:MFCD00000354
  • Molecular Weight:138.14 g/mol
  • Kokend Pt:196 °C
  • Dichtheid:1,138 g/cm³ (25 °C)
  • Flits Pt:71 °C
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:206-960-9
Specificaties

Specificaties

  • Verpakking:
    Bottle
  • VPE:
    1 kg

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.