Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Selenium dioxide 99,9%

Bottle, 100 g
Catalogus # APOSIN3175-100G
Leverancier:   Apollo Scientific
Selenium dioxide 99,9%
Catalogus # APOSIN3175-100G
Leverancier:  Apollo Scientific
CAS-nummer:  7446-08-4
Selenium dioxide 99,9%

Bottle, 100 g

CAS-nummer:  7446-08-4
Price
304,00 €
Log in voor beschikbaarheid
1 * 100 g
Product Details & Documents
  • CAS-nummer:7446-08-4
  • Formule:SeO₂
  • MDL-nummer:MFCD00003562
  • Molecular Weight:110.96 g/mol
  • Smelten Pt:340…350 °C (sublimatie)
  • Dichtheid:3,95 g/cm³ (25 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • UN:3283
  • ADR:6.1,III
Specificaties

Specificaties

  • Verpakking:
    Bottle
  • VPE:
    100 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.