Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Bismuth(III) carbonate basic 99.99%

Bismuth(III) carbonate basic 99.99%

Bottle
Catalogus # APOSIN1239-100G
Leverancier:   Apollo Scientific
Bismuth(III) carbonate basic 99.99%
Catalogus # APOSIN1239-100G
Leverancier:  Apollo Scientific
CAS-nummer:  5892-10-4
Bismuth(III) carbonate basic 99.99%

Bottle

CAS-nummer:  5892-10-4
Price
348,00 €
Log in voor beschikbaarheid
1 * 100 g
Product Details & Documents
Synoniemen:  Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
  • CAS-nummer:5892-10-4
  • Formule:(BiO)₂CO₃
  • MDL-nummer:MFCD00010877
  • Molecular Weight:509.97 g/mol
  • Dichtheid:6,86 g/cm³ (25 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:227-567-9
Specificaties

Specificaties

  • Verpakking:
    Bottle

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.