Orders LinkContactUsLinkComponent
γ-Cyclodextrine
γ-Cyclodextrine
Bottle, 100 g
Catalog # APOSBIM1016-100G
Irritant
Supplier:   Apollo Scientific
γ-Cyclodextrine
Catalog # APOSBIM1016-100G
Supplier:  Apollo Scientific
CAS Number:  17465-86-0
γ-Cyclodextrine
Bottle, 100 g
CAS Number:  17465-86-0
Price
229,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
  • CAS Number:17465-86-0
  • Formula:C₄₈H₈₀O₄₀
  • MDL Number:MFCD00009595
  • Molecular Weight:1297.1248 g/mol
  • Storage Temperature:Kamertemperatuur
Specifications
Specifications
  • Verpakking:
    Bottle
  • VPE:
    100 g
Specifications