Tool carts
Gereedschapswagen met 8 laden (2 grote, 2 middelgrote en 4 kleine), veiligheidsslot en cleanroomwielen met rem., , 900×600×1193 mm
Catalogus # PALBTOOLCART-8-316
Leverancier: Palbam
Ga naar:
Product Details & Documents
Tool cart in stainless steel 304 with electropolished finish.
- Five fully extendable drawers
- One lock which secures all the drawers
- Top ledge to prevent tools/parts falling
- Cleanroom castors with brakes
Documents
Palbam Class - Catalog 2018
Product documentatie
Palbam Class Cleanroom Furniture
Video 's
Specificaties
- Omschrijving:Gereedschapswagen met 8 laden (2 grote, 2 middelgrote en 4 kleine), veiligheidsslot en cleanroomwielen met rem.
- Seq:3
- b×d×h:900×600×1193 mm
- Cat. No.:PALBTOOLCART-8-316