Accessories for U-D master
Sample insert, stainless steel, 1,8 ml
Catalog # SAMP1230T-18
Supplier: Sampling Systems
Jump to:
Product Details & Documents
Specifications
- Omschrijving:Sample insert, stainless steel, 1,8 ml
- Seq:19