Microplates, 384-well, non binding
384-well microplate, Small Volume™, HiBase, non binding, flat bottom, wit
Catalogus # GREI784904_40
Leverancier: Greiner Bio-One
Microplates, 384-well, non binding
Catalogus # GREI784904_40
Leverancier: Greiner Bio-One
Ga naar:
Product Details & Documents
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
All plates are free from RNase, DNase, and pyrogens, and are also free of DNA polymerase inhibitors.
Specificaties
- Kleur:wit
- Omschrijving:384-well microplate, Small Volume™, HiBase, non binding, flat bottom
- Seq:07
- Cat. No.:GREI784904_40
- Milieu certificering:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Lage impact op de fabricage