Orders LinkContactUsLinkComponent
96-Well plate sample control system, U-2D™
96-Well plate sample control system, U-2D™
Inserts, clear RSA-pro glass, 1 ml, for use with U-2D™ rack
Catalogus # MICS95296-I-1MLMR
Leverancier:   MICROSOLV
96-Well plate sample control system, U-2D™
Catalogus # MICS95296-I-1MLMR
Leverancier:  MICROSOLV
96-Well plate sample control system, U-2D™
96-Well plate sample control system, U-2D™
Inserts, clear RSA-pro glass, 1 ml, for use with U-2D™ rack
Price
207,00 €
Log in voor beschikbaarheid
1 * 100 ST
Product Details & Documents
The U-2D™ universal two dimensional 96-well plate system offer users unprecedented sample management control compared to current glass lined or plastic well plates for sample preparation or chromatography.
Easily push up the insert from the bottom and remove it from the rack. These inserts can then be placed into any MicroSolv 9 mm screw top autosampler vial for safe storage, transport or for a second chromatography dimension, or to another instrument platform.
Ordering information: Base and sealing mats sold separately.
Specificaties
  • Omschrijving:
    Inserts, clear RSA-pro glass, 1 ml, for use with U-2D™ rack
  • Seq:
    04
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Paddles for U-2D™ sealing film
    Paddles for U-2D™ sealing film
    MICS95296-PDL-PK
    Log in voor beschikbaarheid
    1 * 5 ST - 17,10 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Roller for U-2D™ sealing film
    Roller for U-2D™ sealing film
    MICS95296-ROLLER
    Log in voor beschikbaarheid
    1 * 1 ST - 65,60 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • U-2D™ plate, 350µL, with rack and base only
    U-2D™ plate, 350µL, with rack and base only
    MICS95296-U2D
    Log in voor beschikbaarheid
    1 * 1 ST - 116,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Zone free sealing film for U-2D™ plates
    Zone free sealing film for U-2D™ plates
    MICS95296-ZFF-PK
    Log in voor beschikbaarheid
    1 * 10 ST - 54,50 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Zone free sealing film for U-2D™ plates
    Zone free sealing film for U-2D™ plates
    MICS95296-ZFF-CS
    Log in voor beschikbaarheid
    1 * 50 ST - 228,00 €