96-Well plate sample control system, U-2D™
U-2D™ rack with 96 well plate, 500 µl preloaded inserts, base and chemical resistant pre-slit mat
Catalog # MICS95296-500-3R
Supplier: MICROSOLV
Jump to:
Product Details & Documents
The U-2D™ universal two dimensional 96-well plate system offer users unprecedented sample management control compared to current glass lined or plastic well plates for sample preparation or chromatography.
Easily push up the insert from the bottom and remove it from the rack. These inserts can then be placed into any MicroSolv 9 mm screw top autosampler vial for safe storage, transport or for a second chromatography dimension, or to another instrument platform.
Ordering information: Base and sealing mats sold separately.
Specifications
- Omschrijving:U-2D™ rack with 96 well plate, 500 µl preloaded inserts, base and chemical resistant pre-slit mat
- Seq:23
Accessories
Product Information
- Catalog #OmschrijvingAvailabilityPrice
Product Information
- Catalog #OmschrijvingAvailabilityPrice
Product Information
- Catalog #OmschrijvingAvailabilityPrice
- MICS95296-U2DU-2D™ plate, 350µL, with rack and base onlyLog in for availability1 * 1 ST - 116,00 €
Product Information
- Catalog #OmschrijvingAvailabilityPrice
- MICS95296-ZFF-PKZone free sealing film for U-2D™ platesLog in for availability1 * 10 ST - 54,50 €
Product Information
- Catalog #OmschrijvingAvailabilityPrice
- MICS95296-ZFF-CSZone free sealing film for U-2D™ platesLog in for availability1 * 50 ST - 228,00 €