Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Diverters for ceiling leaks

Diverters for ceiling leaks

Diverter with magnets, 1524×1524 mm
Catalogus # 119-0299
Leverancier:   JUSTRITE MANUFACTURING
Diverters for ceiling leaks
Catalogus # 119-0299
Leverancier:  JUSTRITE MANUFACTURING
Diverters for ceiling leaks
Diverters for ceiling leaks

Diverter with magnets, 1524×1524 mm

Price
496,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

These ceiling leak diverters redirect roof or pipe leaks to prevent from water damages. The durable, puncture resistant and watertight PVC-coated yellow fabric is designed to channel overhead pipe and ceiling leaks to containers or floor drains.

  • Available with high strength magnets for steel rafter attachment, or grommets for bungee or zip tie attachment
  • Constructed with cinched corners for quick and easy installation
  • Simply connect to a standard garden hose to drain water safely away

Flow rate: 12,5 L/min

Specificaties
  • Omschrijving:
    Diverter with magnets
  • l×b:
    1524×1524 mm