Connecting adapters, with parallel necks
, 18,8/26 NS, 18,8/26 NS
Catalogus # NEUB0900-10-191919
Leverancier: NEUBERT VOLUME GLASSWAERE
Connecting adapters, with parallel necks
Catalogus # NEUB0900-10-191919
Leverancier: NEUBERT VOLUME GLASSWAERE
Ga naar:
Product Details & Documents
Two necked adapters made of borosilicate glass 3.3
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Specificaties
- Afmetingen kern:18,8/26 NS
- Huls:18,8/26 NS
- Seq:01