Orders LinkContactUsLinkComponent
Connecting adapters, with parallel necks
Connecting adapters, with parallel necks
, 29/32 NS, 29/32 NS
Catalogus # NEUB0900-10-292929
Connecting adapters, with parallel necks
Catalogus # NEUB0900-10-292929
Connecting adapters, with parallel necks
, 29/32 NS, 29/32 NS
Price
90,40 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Two necked adapters made of borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Specificaties
  • Afmetingen kern:
    29/32 NS
  • Huls:
    29/32 NS
  • Seq:
    03