Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-10

Mouse Recombinant IL-10

10 µG
Catalogus # STMC78079
Marketsource
Leverancier:   Stemcell Technologies
Mouse Recombinant IL-10
Catalogus # STMC78079
Leverancier:  Stemcell Technologies
Marketsource
CAS-nummer:  
Mouse Recombinant IL-10
Mouse Recombinant IL-10

10 µG

CAS-nummer:  
Price
190,00 €
Log in voor beschikbaarheid
1 * 1 EA
Product Details & Documents

Interleukin 10 (IL-10) is the founding member of the IL-10 family of class II cytokines. All of the IL-10 cytokine family members have a four-helix bundle consisting of α-helical folds. Upon binding to its receptor, IL-10 activates signaling through JAK1 and STAT3. It is produced by dendritic cells, macrophages, and CD4+ T regulatory cells, as well as mast cells, NK cells, neutrophils, and regulatory B cells, under specific stimulating conditions (Saraiva and O'Garra). IL-10 can inhibit the activation of certain immune cells while it promotes the function of B cells, and facilitate healing process. Specifically, this cytokine is important for the function of T regulatory cells as it is a potent suppressor of effector T cell proliferation and cytokine production. Also, IL-10 produced by a subset of macrophages inhibits activation and production of pro-inflammatory cytokines by neighboring macrophages, thus allowing a level of self-regulation. IL-10 enhances B cell proliferation, immunoglobulin secretion, and class II MHC expression (Ouyang et al.).

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specificaties

Specificaties

  • VPE:
    10 µG
  • Cat. No.:
    STMC78079
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-10
  • Species:
    Mouse
  • Zuiverheid:
    ≥95 %

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.