Protease Inhibitor Tablets contain broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation during cell lysis and protein extraction experiments. 1 * 1 EA
Catalogus # PIERA32963
Leverancier: Thermo Fisher Scientific
Frequently Bought Together