ContactUsLinkComponent
Thermo Scientific Pierce Protease Inhibitor Mini Tablets, EDTA-free, contain broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation during cell lysis and protein extraction experiments. 1 * 1 EA
Catalog # PIERA32955
undefined
Thermo Scientific Pierce Protease Inhibitor Mini Tablets, EDTA-free, contain broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation during cell lysis and protein extraction experiments. 1 * 1 EA
Catalog # PIERA32955
Frequently Bought Together