Protease and Phosphatase Inhibitor Mini Tablets, EDTA-free, contain both protease inhibitors and phosphatase inhibitors to provide broad-spectrum complete protection from dephosphorylation and proteolytic degradation. 1 * 1 EA
Catalogus # PIERA32961
Leverancier: Thermo Fisher Scientific
Frequently Bought Together