Calibration reference materials, alloys
Perkalloy, three 50,8 mm pieces (596 °C), 1
Catalogus # PERCN5320114
Leverancier: PerkinElmer
Calibration reference materials, alloys
Catalogus # PERCN5320114
Leverancier: PerkinElmer
CAS-nummer:
Ga naar:
Product Details & Documents
Specificaties
Specificaties
- VPE:1
- Omschrijving:Perkalloy, three 50,8 mm pieces (596 °C)
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



