Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Calibration reference materials, alloys

Calibration reference materials, alloys

Nickel, three 50,8 mm pieces (354 °C), 1
Catalogus # PERCN5320113
Leverancier:   PerkinElmer
Calibration reference materials, alloys
Catalogus # PERCN5320113
Leverancier:  PerkinElmer
CAS-nummer:  
Calibration reference materials, alloys

Nickel, three 50,8 mm pieces (354 °C), 1

CAS-nummer:  
Price
216,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties

Specificaties

  • VPE:
    1
  • Omschrijving:
    Nickel, three 50,8 mm pieces (354 °C)

Specificaties