Calibration reference materials, alloys
Alumel, three 50,8 mm pieces (154 °C), 1
Catalogus # PERCN5320110
Leverancier: PerkinElmer
Calibration reference materials, alloys
Catalogus # PERCN5320110
Leverancier: PerkinElmer
CAS-nummer:
Ga naar:
Product Details & Documents
Specificaties
Specificaties
- VPE:1
- Omschrijving:Alumel, three 50,8 mm pieces (154 °C)



