Multi-packs, Proline® Plus
3+1-pack, incl. three single channel pipettes (0,5 - 10 µl, 20 - 200 µl, 100 - 1000 µl), one 8-channel pipette (30 - 300 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 1× 0,5 - 200 µl, 1× 5 - 350 µl, 1× 10 - 1000 µl) and a linear stand
N° de catalogue 613-5738
Fournisseur: Sartorius
Multi-packs, Proline® Plus
N° de catalogue 613-5738
Fournisseur: Sartorius
Aller à :
Product Details & Documents
These Multipacks combine a set of Proline® Plus mechanical pipettes with matching tip trays and useful accessories. They offer an opportunity to test and get started with Proline® Plus mechanical pipettes.
- Ergonomic design with light pipetting forces, comfortable handle and finger support for reduced risk of strain injuries
- Minimised risk of contamination with Safe-Cone Filters and full autoclavability
- Highly durable with strengthened structure also for heavier use
It is the widest range of pipettes for various users and applications.
Livraison: All packs include a calibration tool that also acts as a tube opener, as well as the required instructions.
Services: Voor erkende kalibratie onder ISO8655 en ISO17025, onderhoud en reparatie mail naar [email protected]
Spécifications
- Omschrijving:3+1-pack, incl. three single channel pipettes (0,5 - 10 µl, 20 - 200 µl, 100 - 1000 µl), one 8-channel pipette (30 - 300 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 1× 0,5 - 200 µl, 1× 5 - 350 µl, 1× 10 - 1000 µl) and a linear stand
- Seq:04
- Cat. No.:613-5738
- Milieu certificering:Renewable energy certificate; ISO 14001
- Environmentally Preferable:Lage impact op de fabricage