Multipacks, Tacta®
The Tacta® Multipacks offer an opportunity to test and get started with Tacta® mechanical pipettes. Tacta® mechanical pipette offers excellent performance and safety in manual pipetting. It is designed to maintain high accuracy and precision in repetitive and long-lasting pipetting series.
- Reliable results, even over lengthy pipetting periods
- Controlled and smooth tip ejection
- The Optilock® system provides flexibility for volume adjustment and locking
There are different multipack options to choose from ranging from a set of 3 to a set of 5 pipettes.
Ordering information: All sets come with a linear stand for the safe storage of your pipettes as well as suitable racked or bulk-packed Optifit tips.
Services: Voor erkende kalibratie onder ISO8655 en ISO17025, onderhoud en reparatie mail naar [email protected]
- Omschrijving:3+1-pack, incl. three single channel pipettes (0,5 - 10 / 20 - 200 / 100 - 1000 µl) and one 8-channel pipette (30 - 300 µl), Optifit tip tray boxes (0,1 - 10 / 0,5 - 200 / 5 - 350 / 10 - 1000 μl; 96 tips each) and one linear stand
- Seq:07
- Cat. No.:613-5632
- Milieu certificering:Renewable energy certificate; ISO 14001
- Environmentally Preferable:Lage impact op de fabricage