Orders LinkContactUsLinkComponent
Accessories
Telescopic feed tubing, FEP, 150 - 255 mm, for dispensers up to 10 ml
Catalogus # SOCO1.525.352
Leverancier:   Socorex Isba
Accessories
Catalogus # SOCO1.525.352
Leverancier:  Socorex Isba
Accessories
Accessories
Telescopic feed tubing, FEP, 150 - 255 mm, for dispensers up to 10 ml
Price
33,50 €
Log in voor beschikbaarheid
1 * 1 ST
Specificaties
  • Omschrijving:
    Telescopic feed tubing, FEP, 150 - 255 mm, for dispensers up to 10 ml
  • Seq:
    20