Affinity chromatography columns, HiTrap™ IXSelect
HiTrap™ IXSelect, PP, 5 ml
Catalogus # GEHE17-3714-12
Leverancier: Cytiva
Ga naar:
Product Details & Documents
HiTrap™ IXSelect columns are prepacked with IXSelect resin for screening and small-scale protein purifications using affinity chromatography.
- IXSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams
Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Aanvullende productdetails
Specificaties
- Capaciteit:5 ml
- Omschrijving:HiTrap™ IXSelect
- Kolombehuizing:PP
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.