Orders LinkContactUsLinkComponent
Spray chambers and brackets
Spray chambers and brackets
PuraFlex transfer tube for IsoMist used with 5100 ICP-OES, required to connect aerosol outlet from IsoMist temperature-controlled spray chamber to easy-fit torch
Catalogus # HEWLG8010-68011
Leverancier:   Agilent
Spray chambers and brackets
Catalogus # HEWLG8010-68011
Leverancier:  Agilent
Spray chambers and brackets
Spray chambers and brackets
PuraFlex transfer tube for IsoMist used with 5100 ICP-OES, required to connect aerosol outlet from IsoMist temperature-controlled spray chamber to easy-fit torch
Price
495,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
These ICP-OES series are compatible with a range of accessories and supplies designed to extend the capabilities of the instruments.
Single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout.
Double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Sturman-Masters spray chamber is inert and minimizes interferences and improves stability with high TDS samples.
Specificaties
  • Omschrijving:
    PuraFlex transfer tube for IsoMist used with 5100 ICP-OES, required to connect aerosol outlet from IsoMist temperature-controlled spray chamber to easy-fit torch
  • Seq:
    06
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.