Inlaat liners
Liner, ultra inert deactivation, straight, 0.75 mm
Catalogus # HEWL5190-7045
Leverancier: Agilent
Ga naar:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Specificaties
- Ø inw.:0.75 mm
- Omschrijving:Liner, ultra inert deactivation, straight
- Seq:10