Instrument trays
490×320×15 mm
Catalogus # SAMPA674-490
Leverancier: Sampling Systems
Ga naar:
Product Details & Documents
Made from 304 stainless steel.
- Good chemical resistance, very good temperature resistance
- Crevice free design
- High quality polished finish
Ideal for general laboratory use as well as for carrying surgical instruments, sampling tools etc.
Documents
SAMPA674-215_TDS_EN
Technische fiche
Specificaties
- Seq:04
- b×d×h:490×320×15 mm