Orders LinkContactUsLinkComponent
Instrument trays
Instrument trays
215×160×18 mm
Catalogus # SAMPA674-215
Leverancier:   Sampling Systems
Instrument trays
Catalogus # SAMPA674-215
Leverancier:  Sampling Systems
Instrument trays
Instrument trays
215×160×18 mm
Price
24,90 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Made from 304 stainless steel.
  • Good chemical resistance, very good temperature resistance
  • Crevice free design
  • High quality polished finish
Ideal for general laboratory use as well as for carrying surgical instruments, sampling tools etc.
Documents
Specificaties
  • Seq:
    01
  • b×d×h:
    215×160×18 mm