α-Cyclodextrin ≥98.0% (by HPLC)
100 g
Catalogus # TCIAC0776-100G
Leverancier: TCI
Ga naar:
Product Details & Documents
- CAS-nummer:10016-20-3
- Formule:C₃₆H₆₀O₃₀
- MDL-nummer:MFCD00078207
- Molecular Weight:972.84 g/mol
- Merck index:14,02718
- EINECS:233-007-4
Specificaties
Specificatie testresultaten
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+146.0 to +153.0 deg (C=1, H2O) (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:max. 10.0 %
Specificaties
- VPE:100 g
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



