Orders LinkContactUsLinkComponent
MultiDuster mopping systems
MultiDuster mopping systems
MultiDuster maxi MicroPlus sleeve (CC), 465 mm
Catalog # VILE151719
Supplier:   FHP VILEDA SCS
MultiDuster mopping systems
Catalog # VILE151719
Supplier:  FHP VILEDA SCS
MultiDuster mopping systems
MultiDuster mopping systems
MultiDuster maxi MicroPlus sleeve (CC), 465 mm
Price
508,00 €
Log in for availability
1 * 20 ST
Product Details & Documents
MultiDuster has a unique mechanism that brings significant benefits like hygienic cleaning of high and inaccessible areas.
  • Easy to wipe clean
  • Doesn’t harbour bacteria
  • Fits to handle in all bent positions
  • Double sided sleeves made of microfibre
  • Returns to straight position
  • Fits telescopic handles
MultiDuster Maxi easily cleans pipes and ledges up to 5 m high. For low areas the MultiDuster Mini is the perfect hand-held solution. Both Maxi and Mini have slim designs, for easy cleaning behind tight spaces such as radiators, between glass partitions, around pipes and many more.
It is suitable for health care industries, general building and food industries.
Certifications: MicroPlus sleeves are Nordic Swan certified.
Specifications
  • Omschrijving:
    MultiDuster maxi MicroPlus sleeve (CC)
  • Lengte:
    465 mm
  • Seq:
    31