Orders LinkContactUsLinkComponent
Thread adapter, GL PTFE , BOLA
Thread adapter, GL PTFE , BOLA
With 2 GL threads, GL14 (from male thread) and GL25 (to male thread), 8,5 mm
Catalogus # BOHLD537-08
Leverancier:   Bohlender
Thread adapter, GL PTFE , BOLA
Catalogus # BOHLD537-08
Leverancier:  Bohlender
Thread adapter, GL PTFE , BOLA
Thread adapter, GL PTFE , BOLA
With 2 GL threads, GL14 (from male thread) and GL25 (to male thread), 8,5 mm
Price
69,50 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Straight transition thread adapter made of PTFE with two GL-threaded connections.
  • Chemical resistant
  • Vacuum suitable
  • Temperature resistant from –200 to +250 °C
Specificaties
  • Boring Ø:
    8,5 mm
  • Omschrijving:
    With 2 GL threads, GL14 (from male thread) and GL25 (to male thread)
  • Seq:
    02
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.