Orders LinkContactUsLinkComponent
Cryoboxes, PP
7×7, groen, 133×133×50/75 mm
Catalog # RATI5320313
Supplier:   RATIOLAB
Cryoboxes, PP
Catalog # RATI5320313
Supplier:  RATIOLAB
Cryoboxes, PP
Cryoboxes, PP
7×7, groen, 133×133×50/75 mm
Price
41,40 €
Log in for availability
1 * 5 ST
Product Details & Documents
Boxes made of high quality PP.
  • Autoclavable
  • Temperature-resistant from –90 to +121 °C
  • Numerically coded
  • Very good chemical resistance
With an numerical coding and corresponding vents for draining condensed water. The boxes are adjustable in height.
Specifications
  • Compartimenten:
    7×7
  • Uitw. b×d×h:
    133×133×50/75 mm
  • Kleur:
    groen
  • Seq:
    01