Orders LinkContactUsLinkComponent
Adapters with stopcock and socket
Adapters with stopcock and socket
24/29 NS, 3 mm
Catalogus # BIBBMF14/3/SC
Leverancier:   SciLabware
Adapters with stopcock and socket
Catalogus # BIBBMF14/3/SC
Leverancier:  SciLabware
Adapters with stopcock and socket
Adapters with stopcock and socket
24/29 NS, 3 mm
Price
573,00 €
Log in voor beschikbaarheid
1 * 5 ST
Product Details & Documents
PYREX®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With GP Rotaflo® PTFE stopcock providing fine flow control and positive closure. Detachable plastic screwthread connectors allow rubber hose etc. to be fitted easily and safely .
Specificaties
  • Boring Ø:
    3 mm
  • Huls:
    24/29 NS