ContactUsLinkComponent
[EN]SAVANA PTFE-HPX 0.2UM 10 IN C-0 E-FK 1 * 1 ST
Catalog # ENTESV0G01P01
Supplier:  ENTEGRIS
[EN]SAVANA PTFE-HPX 0.2UM 10 IN C-0 E-FK 1 * 1 ST
Catalog # ENTESV0G01P01
Supplier:  ENTEGRIS
Frequently Bought Together