Druppelflessen, Lucas
10 ml
Catalogus # MUEL14.196.02
Leverancier: MUELLER KREMPEL
Ga naar:
Product Details & Documents
Amber glass, without closure.
- Good chemical and physical properties
- Use in applications that do not involve high thermal stresses
- Please order screw caps and drop dispensers separately
Bruingekleurd ter bescherming van lichtgevoelige materialen.
Specificaties
- Capaciteit:10 ml
- Seq:03