SODIUM HYDROXIDE 0.01 N IN ETHANOL 1 * 1 L
Catalogus # PRLS89630.290
Leverancier: PROLABO CMP
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
VWR®, gezichtstissues
Leverancier: VWR Collection
pH-teststrips, MQuant®, Supelco®
Leverancier: Merck
Testsets, ammoniumtest, voor Spectroquant®, Supelco®
Leverancier: Merck
pH-indicatiestrips, MQuant®, Supelco®
Leverancier: Merck
Natriumhydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Leverancier: VWR Chemicals
Testsets, Nitriettest voor Spectroquant®, Supelco®
Leverancier: Merck
Testsets COD-cuvettentest voor Spectroquant®, Supelco®
Leverancier: Merck
VWR® Rota®, pH indicator paper, 1 to 14
Leverancier: VWR Chemicals
Natriumchloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Leverancier: Merck
Natriumhydroxide 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
Leverancier: VWR Chemicals
Teststrips, nitraattest, MQuant™, Supelco®
Leverancier: Merck
Teststrips, peroxidetest, MQuant™, Supelco®
Leverancier: Merck
Test strips, peroxide, Dosatest
Leverancier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Leverancier: VWR Chemicals
Test kits, photometric, TOC, LCK cuvette tests
Leverancier: Hach