Orders LinkContactUsLinkComponent
Comparator kits for water testing
Comparator kits for water testing
Ozone coloTest Kit, AccuVac® Color Disc, 1 KIT
Catalogus # HACH2518050
Leverancier:   Hach
Comparator kits for water testing
Catalogus # HACH2518050
Leverancier:  Hach
CAS-nummer:  
Comparator kits for water testing
Comparator kits for water testing
Ozone coloTest Kit, AccuVac® Color Disc, 1 KIT
CAS-nummer:  
Price
202,00 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents
Qualitative results using traditional colour comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specificaties
Specificaties
  • VPE:
    1 KIT
  • Omschrijving:
    Ozone coloTest Kit, AccuVac® Color Disc
Specificaties
Accessories
Productinformatie
  • Catalogus #
    VPE
    Beschikbaarheid
    Price
  • Ammonium molybdate reagent for phosphate determinations, 100 ml
    Ammonium molybdate reagent for phosphate determinations, 100 ml
    HACH11032H
    1
    Log in voor beschikbaarheid
    1 * 1 ST - 27,90 €