Orders LinkContactUsLinkComponent
Filling solutions for combined/reference electrodes
Filling solutions for combined/reference electrodes
K2SO4 solution, saturated, 30 ml, 30 mL
Catalogus # HACHS21M007
Leverancier:   Hach
Filling solutions for combined/reference electrodes
Catalogus # HACHS21M007
Leverancier:  Hach
CAS-nummer:  
Filling solutions for combined/reference electrodes
Filling solutions for combined/reference electrodes
K2SO4 solution, saturated, 30 ml, 30 mL
CAS-nummer:  
Price
18,20 €
Log in voor beschikbaarheid
1 * 30 mL
Product Details & Documents
Filling and storage solutions for combined and reference electrodes.
Documents
Specificaties
Specificaties
  • VPE:
    30 mL
  • Omschrijving:
    K<sub>2</sub>SO<sub>4 </sub>solution, saturated, 30 ml