Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Petrischalen

Petrischalen

With 3 vents, 150 mm, 15 mm, Aseptisch, 8×20/bag
Catalogus # ANICP150V000
Leverancier:   Anicrin
Petrischalen
Catalogus # ANICP150V000
Leverancier:  Anicrin
Petrischalen
Petrischalen

With 3 vents, 150 mm, 15 mm, Aseptisch, 8×20/bag

Price
213,00 €
Log in voor beschikbaarheid
1 * 220 ST
Specificaties
  • Ø:
    150 mm
  • Verpakt:
    8×20/bag
  • Steriliteit:
    Aseptisch
  • Uitvoering:
    With 3 vents
  • Hoogte:
    15 mm
  • Seq:
    13
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.